Learn More
(R/S)-N-Deacetyl Colchicine, TRC
Specifications
Specifications
| Chemical Name or Material | (R/S)-N-Deacetyl Colchicine |
| CAS | 102491-73-6 |
| Molecular Formula | C20H23NO5 |
| Quantity | 10 mg |
| Synonym | 7-Amino-6,7-dihydro-1,2,3,10-tetramethoxy-benzo[a]heptalen-9(5H)-one, (+/-)-7-Deacetylcolchicine |
| InChI Formula | InChI=1S/C20H23NO5/c1-23-16-8-6-12-13(10-15(16)22)14(21)7-5-11-9-17(24-2)19(25-3)20(26-4)18(11)12/h6,8-10,14H,5,7,21H2,1-4H3 |
| SMILES | COC1=CC=C2C(=CC1=O)C(N)CCc3cc(OC)c(OC)c(OC)c23 |
| IUPAC Name | 7-amino-1,2,3,10-tetramethoxy-6,7-dihydro-5H-benzo[a]heptalen-9-one |
| Molecular Weight (g/mol) | 357.4 |
| Formula Weight | 357.1576 |