Learn More
(R,R)-Tartaric Acid Monomethyl Ester Diacetate, TRC
Specifications
Specifications
| Chemical Name or Material | (R,R)-Tartaric Acid Monomethyl Ester Diacetate |
| CAS | 23294-83-9 |
| Molecular Formula | C9H12O8 |
| Quantity | 2.5 g |
| Synonym | (2R,3R)-2,3-Bis(acetyloxy)-butanedioic Acid Monomethyl Ester, [R-(R*,R*)]-2,3-Bis(acetyloxy)-butanedioic Acid Monomethyl Ester |
| InChI Formula | InChI=1S/C9H12O8/c1-4(10)16-6(8(12)13)7(9(14)15-3)17-5(2)11/h6-7H,1-3H3,(H,12,13)/t6-,7-/m1/s1 |
| SMILES | COC(=O)[C@H](OC(=O)C)[C@@H](OC(=O)C)C(=O)O |
| IUPAC Name | (2R,3R)-2,3-diacetyloxy-4-methoxy-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 248.19 |
| Formula Weight | 248.0532 |