missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-Sertraline, TRC

Product Code. 30428360
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428360 2.5 mg 2.5mg
30411690 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428360 Supplier Toronto Research Chemicals Supplier No. TRCS2799602.5MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79617-98-4
Molecular Formula C17 H17 Cl2 N
Molecular Weight (g/mol) 306.2296
Synonym (1R,4R)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine, (1R-cis)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-Naphthalenamine, (1R,4R)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine, CP 51973
IUPAC Name (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine
SMILES CN[C@H]1CC[C@@H](c2ccc(Cl)c(Cl)c2)c3ccccc13

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H17Cl2N/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11/h2-6,8,10,12,17,20H,7,9H2,1H3/t12-,17-/m0/s1
Formula Weight 305.0738
Chemical Name or Material (R,R)-Sertraline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.