Learn More
R&D Systems™ Quantikine Immunoassay Control Set 733 for Human FLRG
Product Code. 18749437
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18749437 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18749437 Supplier R&D Systems™ Supplier No. QC91
  • ONLINE EXCLUSIVE PRICE
    210.00 EUR
    198.45 EUR / Pack of 1
    Save 11.55 EUR
    5% Off
 Restricted Item
Estimated Shipment: 10-11-2026
For use with catalog number DFLRG0
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Follistatin-related Gene Protein (FLRG)
Gene ID (Entrez) 10272
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human