Learn More
(R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide, TRC
Product Code. 30473052
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473052 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30473052 Supplier Toronto Research Chemicals Supplier No. TRCN378485200MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propanamide
CAS 1005450-55-4
Molecular Formula C22 H20 F3 N O
Quantity 200 mg
Synonym Benzenepropanamide','N-[(1R)-1-(1-naphthalenyl)ethyl]-3-(trifluoromethyl)-, N-[(1R)-1-(1-Naphthalenyl)ethyl]-3-(trifluoromethyl)benzenepropanamide, N-[1-(R)-(1-Naphthyl)ethyl]-3-[3-trifluoromethylphenyl]propanamide
InChI Formula InChI=1S/C22H20F3NO/c1-15(19-11-5-8-17-7-2-3-10-20(17)19)26-21(27)13-12-16-6-4-9-18(14-16)22(23,24)25/h2-11,14-15H,12-13H2,1H3,(H,26,27)/t15-/m1/s1
SMILES C[C@@H](NC(=O)CCc1cccc(c1)C(F)(F)F)c2cccc3ccccc23
IUPAC Name N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]propanamide
Molecular Weight (g/mol) 371.4
Formula Weight 371.1497