Learn More
(R)-(+)-Limonene, Sigma-Aldrich™
Product Code. 15890391
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15890391 100 mL 100mL
15800401 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15890391 Supplier Sigma Aldrich Supplier No. 8184070100
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5989-27-5
Grade Research
Molecular Weight (g/mol) 136.23
Specifications
Melting Point -74°C
Density 0.84 g/cm3 at 20°C
Boiling Point 177°C to 178°C/1013 hPa
MDL Number MFCD00062991
Quantity 100 mL
Synonym (R)-(+)-Limonene, (+)-p-Mentha-1,8-diene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, (R)-(+)-Limonene