missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Ibuprofen, MedChemExpress

Product Code. 16421346
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421346 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16421346 Supplier Medchem Express Supplier No. HY78131B
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (R)-(-)-Ibuprofen is the R enantiomer of Ibuprofen, inactive on COX, inhibits NF-κB activation; (R)-(-)-Ibuprofen exhibits anti-inflammatory and antinociceptive effects.

(R)-(-)-Ibuprofen is the R enantiomer of Ibuprofen, inactive on COX, inhibits NF-κB activation

Specifications

Chemical Name or Material (R)-(-)-Ibuprofen
Physical Form Solid
CAS 51146-57-7
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C13H18O2
For Use With (Application) COVID-19-immunoregulation
Synonym (R)-Ibuprofen
Solubility Information DMSO : ≥ 100 mg/mL (484.78 mM) ∣H2O : 0.67 mg/mL (3.25 mM; Need ultrasonic)
SMILES C[C@H](C1=CC=C(CC(C)C)C=C1)C(O)=O
Molecular Weight (g/mol) 206.28
Formula Weight 206.28
Percent Purity 98.49%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.