missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human PTP alpha/PTPRA (NP_543030) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Human PTP alpha/PTPRA (NP_543030) VersaClone cDNA clone contains the complete ORF for PTP alpha/PTPRA, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PTP alpha/PTPRA insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
| Gene | PTP alpha/PTPRA |
| Accession Number | NP_543030 |
| Gene ID (Entrez) | 5786 |
| Gene Alias | EC 3.1.3.48, HEPTP, HLPR, LRP, LRPHPTPalpha, protein tyrosine phosphatase, receptor type, A, Protein-tyrosine phosphatase alpha, PTPAreceptor type, alpha polypeptide, PTPase-alpha, PTPLCA-related phosphatase, PTPRA, PTPRL2, receptor-type tyrosine-protein phosphatase alpha, RPTPA, R-PTP-alpha, tyrosine phosphatase alpha |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction