Learn More
R&D Systems™ Human IL-1 RAcP/IL-1 R3 (NP_002173) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Conditionnement:
10µg
Description
The Human IL-1 RAcP/IL-1 R3 (NP_002173) VersaClone cDNA clone contains the complete ORF for IL-1 RAcP/IL-1 R3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-1 RAcP/IL-1 R3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Informations sur la commande
Ships at ambient temperature
Spécification
Spécification
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu