missing translation for 'onlineSavingsMsg'
Learn More

(R)-Elagolix, MedChemExpress

Product Code. 16496546
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496546 100 mg 100mg
16476168 10 mg 10mg
16458767 5 mg 5mg
16452307 50 mg 50mg
16430226 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496546 Supplier Medchem Express Supplier No. HY14789

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elagolix is a highly potent, selective, orally-active, short-duration, non-peptide antagonist of the gonadotropin-releasing hormone receptor (GnRHR) (KD = 54 pM).

Elagolix is a highly potent, selective, orally-active, short-duration, non-peptide antagonist of the gonadotropin-releasing hormone receptor (GnRHR) (KD = 54 pM).

Specifications

Chemical Name or Material (R)-Elagolix
CAS 834153-87-6
Color White
Purity Grade Notes Research
Molecular Formula C32H30F5N3O5
Quantity 100 mg
Synonym NBI-56418
Solubility Information DMSO : ≥ 100 mg/mL (158.33 mM)
SMILES FC1=C(C2=C(C)N(CC3=C(C(F)(F)F)C=CC=C3F)C(N(C[C@](C4=CC=CC=C4)([H])NCCCC(O)=O)C2=O)=O)C=CC=C1OC
Molecular Weight (g/mol) 631.59
Formula Weight 631.59
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.