Learn More
R&D Systems™ TACS Nuclease and Buffer
Description
- TACS-Nuclease has high specific activity, is tolerant to a variety of buffer conditions, and is stable at both 4°C and at -20°C
- TACS-Nuclease Buffer has been optimized for use with TACS-Nuclease for the generation of a positive control for DNA fragmentation characteristic of apoptosis
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 15 μL |
| Product Type | TACS Nuclease and Buffer |