missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OICR 9429

Description
OICR 9429 is a high affinity WDR5 antagonist (Kd = 93 nM; IC50 = 64 nM); disrupts WDR5-MLL interaction. Exhibits selectivity for WDR5 over 22 methyltransferases and a panel of kinases, GPCRs, ion channels, and transporters. Reduces viability of acute myeloid leukemia cells in vitro. Also disrupts MLL1-RbBP5 interaction.
To request the negative control for OICR 9429, please fill out the OICR 0547 request form on the SGC website.
Specifications
Specifications
| Chemical Name or Material | OICR 9429 |
| Synonym | N-[2-(4-Methylpiperazin-1-yl)-5-[3-(morpholin-4-ylmethyl)phenyl]phenyl]-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide |
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in DMSO |
| Target | Additional Lysine Methyltransferase Compounds |
| Molecular Formula | C29H32F3N5O3 |
| Formula Weight | 555.59 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?