missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ OICR 9429
GREENER_CHOICE

Product Code. 18738788
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738788 50 mg 50mg
18756386 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18738788 Supplier Tocris Bioscience™ Supplier No. 5267/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective WDR5 antagonist

OICR 9429 is a high affinity WDR5 antagonist (Kd = 93 nM; IC50 = 64 nM); disrupts WDR5-MLL interaction. Exhibits selectivity for WDR5 over 22 methyltransferases and a panel of kinases, GPCRs, ion channels, and transporters. Reduces viability of acute myeloid leukemia cells in vitro. Also disrupts MLL1-RbBP5 interaction.

To request the negative control for OICR 9429, please fill out the OICR 0547 request form on the SGC website.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material OICR 9429
Synonym N-[2-(4-Methylpiperazin-1-yl)-5-[3-(morpholin-4-ylmethyl)phenyl]phenyl]-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Target Additional Lysine Methyltransferase Compounds
Molecular Formula C29H32F3N5O3
Formula Weight 555.59
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.