Learn More
R&D Systems™ Caspase-2 Inhibitor Z-VDVAD-FMK
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
- Fluoromethyl ketone (FMK)-derivatized peptides are useful tools for studying Caspase activity
- Used in tissue cultures to inhibit apoptosis at final working concentrations of 50nM to 100μM
- Regulated by a variety of stimili, including APAF1, CFLAR/FLIP, NOL3/ARC, IAP, BIRC1/NAIP, BIRC2/cIAP-1, BIRC3/cIAP-2, BIRC4/XIAP, BIRC5/Survivin and BIRC7/Livin
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 mg |
| Product Type | Caspase-2 Inhibitor Z-VDVAD-FMK |
| Molecular Weight (g/mol) | 695Da |
| Inhibitors | Z-VDVAD-FMK |
| Form | Lyophilized |
For research use only. Not for use in diagnostic procedures