missing translation for 'onlineSavingsMsg'
Learn More

R 788 Sodium Salt Hydrate, TRC

Product Code. 30421461
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421461 5 mg 5mg
30421202 10 mg 10mg
30413028 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421461 Supplier Toronto Research Chemicals Supplier No. TRCR0701155MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material R 788 Sodium Salt Hydrate
Molecular Formula C23H24FN6Na2O9P • x(H2O)
Quantity 5 mg
Synonym 6-[[5-Fluoro-2-[(3,4,5-trimethoxyphenyl)amino]-4-pyrimidinyl]amino]-2,2-dimethyl-4-[(phosphonooxy)methyl]-2H-pyrido[3,2-b]-1,4-oxazin-3(4H)-one Sodium Salt Hydrate, Fostamatinib Disodium Hydrate, R 935788 sodium Hydrate, R 788 sodium Hydrate
SMILES FC1=CN=C(NC2=CC(OC)=C(OC)C(OC)=C2)N=C1NC(C=C3)=NC4=C3OC(C)(C)C(N4COP(O[Na])(O[Na])=O)=O.O
Molecular Weight (g/mol) 624.421801
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.