Learn More
(R)-5-Bromo-3-(N-methylpyrrolidin-2-ylmethyl)-1H-indole, TRC
Specifications
Specifications
| Chemical Name or Material | R-5-Bromo-3-(N-methylpyrrolidin-2-ylmethyl)-1H-indole |
| CAS | 143322-57-0 |
| Molecular Formula | C14 H17 Br N2 |
| Quantity | 100 mg |
| Synonym | 1H-Indole, 5-bromo-3-[[(2R)-1-methyl-2-pyrrolidinyl]methyl]-, 1H-Indole, 5-bromo-3-[(1-methyl-2-pyrrolidinyl)methyl]-, (R)-, 5-Bromo-3-[[(2R)-1-methyl-2-pyrrolidinyl]methyl]-1H-indole, (R)-5-Bromo-3-(N-methylpyrrolidin-2-ylmethyl)-1H-indole, 5-Bromo-3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-1H-indole, Eletriptan Related Compound B (USP), (R)-5-Bromo-3-[(1-methylpyrrolidin-2-yl)methyl]-1H-indole |
| InChI Formula | InChI=1S/C14H17BrN2/c1-17-6-2-3-12(17)7-10-9-16-14-5-4-11(15)8-13(10)14/h4-5,8-9,12,16H,2-3,6-7H2,1H3/t12-/m1/s1 |
| SMILES | CN1CCC[C@@H]1Cc2c[nH]c3ccc(Br)cc23 |
| IUPAC Name | 5-bromo-3-[[(2R)-1-methylpyrrolidin-2-yl]methyl]-1H-indole |
| Molecular Weight (g/mol) | 293.2 |
| Formula Weight | 292.0575 |