Learn More
(R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 98%
Product Code. 15498036
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498036 250 mg 250mg
15469398 1 g 1g
15479398 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15498036 Supplier Thermo Scientific Alfa Aesar Supplier No. H62954.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 114360-56-4 | C24H28N2O6 | 440.496 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 114360-56-4
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.496
MDL Number MFCD00798647
InChI Key LIWKOFAHRLBNMG-HXUWFJFHSA-N
Synonym fmoc-d-dab boc-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, n-a-fmoc-n-g-boc-d-2,4-diaminobutyric acid, 2r-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-t-butyloxycarbonyl-d-2,4-diaminobutyric acid, ambotzfaa1317, fmoc-4-boc-d-2,4-diaminobutyric acid, n-, a-fmoc-n-, a-boc-d-2,4-diaminobutyricacid, n-a-fmoc-n-gamma-boc-d-2,4-diaminobutyric acid, 2r-4-boc-amino-2-fmoc-amino-butanoic acid
PubChem CID 7021129
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Specifications
Color White
Quantity 250 mg
Formula Weight 440.49
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid