Learn More
(R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium Chloride, TRC
Product Code. 30135894
Change view
Click to view available options
Quantity:
250 mg
5 g
25 g
Unit Size:
250mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135894 5 g 5g
17982535 25 g 25g
17951545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135894 Supplier Toronto Research Chemicals Supplier No. TRCC1763855G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6645-46-1
Molecular Formula C7 H16 N O3 . Cl
Molecular Weight (g/mol) 197.66
Synonym 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, chloride, (2R)-, 1-Propanaminium, 3-carboxy-2-hydroxy-N,N,N-trimethyl-, chloride, (R)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, (-)- (8CI), (R)-(-)-Carnitine hydrochloride, (R)-Carnitine hydrochloride, L-(-)-Carnitine hydrochloride, LC 80, Levocarnitine chloride, l-Carnitine chloride, l-Carnitine hydrochloride
IUPAC Name [(2R)-3-carboxy-2-hydroxy-propyl]-trimethyl-ammonium;chloride
SMILES [Cl-].C[N+](C)(C)C[C@H](O)CC(=O)O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C7H15NO3.ClH/c1-8(2,3)5-6(9)4-7(10)11;/h6,9H,4-5H2,1-3H3;1H/t6-;/m1./s1
Formula Weight 197.0819
Chemical Name or Material Levocarnitine Chloride