Learn More
(R)-3-(Boc-amino)piperidine, TRC
Product Code. 30336596
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336596 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336596 Supplier Toronto Research Chemicals Supplier No. TRCB6211351G
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R)-3-(Boc-amino)piperidine
CAS 309956-78-3
Molecular Formula C10H20N2O2
Quantity 1 g
Synonym (3R)-3-Piperidinyl-carbamic Acid 1,1-Dimethylethyl Ester, (3R)-3-[((tert-Butyloxycarbonyl)amino)]piperidine, (R)-3-(tert-Butoxycarbonylamino)piperidine, (R)-3-[(tert-Butoxycarbonyl)amino]piperidine, (R)-3-[(tert-Butyloxycarbonyl)amino]piperidine, (R)-3-[N-(tert-Butoxycarbonyl)amino]piperidine, (R)-3-tert-Butoxycarbonylaminopiperidine, (R)-Piperidin-3-ylcarbamic acid tert-butyl ester, (R)-tert-Butyl (Piperidin-3-yl)carbamate, tert-Butyl (R)-N-(3-Piperidyl)carbamate, tert-Butyl (R)-N-(Piperidin-3-yl)carbamate, tert-Butyl (R)-piperidin-3-ylcarbamate
InChI Formula InChI=1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-5-4-6-11-7-8/h8,11H,4-7H2,1-3H3,(H,12,13)/t8-/m1/s1
SMILES CC(C)(C)OC(=O)N[C@@H]1CCCNC1
IUPAC Name tert-butyl N-[(3R)-piperidin-3-yl]carbamate
Molecular Weight (g/mol) 200.28
Formula Weight 200.1525