Learn More
(R)-3-(Boc-amino)piperidine, 97%
Product Code. p-7035045
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459487 1 g 1g
15469487 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15459487 Supplier Thermo Scientific Alfa Aesar Supplier No. H52749.03
Add to basket
CAS: 309956-78-3 | C10H20N2O2 | 200.282 g/mol
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol and ethanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 309956-78-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03093382
InChI Key WUOQXNWMYLFAHT-MRVPVSSYSA-N
Synonym r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester
PubChem CID 1514172
IUPAC Name tert-butyl N-[(3R)-piperidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1
Specifications
Solubility Information Soluble in methanol and ethanol.
Quantity 1 g
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)piperidine