Learn More
(R)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid, 97%, BLD Pharm™
Product Code. 30949002
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30949002 25 g 25g
30943995 5 g 5g
30942000 100 g 100g
30942798 10 g 10g
30947615 250 mg 250mg
30941586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30949002 Supplier BLD Pharm Supplier No. BD11794725g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 228857-58-7
Molecular Formula C9H15NO4
MDL Number MFCD01320888
Quantity 25 g
SMILES O=C([C@@H]1N(C(OC(C)(C)C)=O)CC1)O
Molecular Weight (g/mol) 201.22
Percent Purity 97%
Chemical Name or Material (R)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid