Learn More
Quinoline-5-sulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Quinoline-5-sulfonyl Chloride |
| CAS | 102878-84-2 |
| Molecular Formula | C9H6ClNO2S |
| Quantity | 100 mg |
| Synonym | 5-Quinolinesulfonyl Chloride |
| InChI Formula | InChI=1S/C9H6ClNO2S/c10-14(12,13)9-5-1-4-8-7(9)3-2-6-11-8/h1-6H |
| SMILES | ClS(=O)(=O)c1cccc2ncccc12 |
| IUPAC Name | quinoline-5-sulfonyl chloride |
| Molecular Weight (g/mol) | 227.67 |
| Formula Weight | 226.9808 |