missing translation for 'onlineSavingsMsg'
Learn More

Quininone, TRC

Product Code. 17971285
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971285 250 mg 250mg
17922915 2.5 g 2.5g
17992505 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971285 Supplier Toronto Research Chemicals Supplier No. TRCQ694400250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-31-1
Molecular Formula C20 H22 N2 O2
Molecular Weight (g/mol) 322.4
Synonym (8α)-6'-Methoxycinchonan-9-one, Quininone (7CI,8CI), ICQ 15
IUPAC Name [(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanone
SMILES COc1ccc2nccc(C(=O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H22N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19H,1,7,9-10,12H2,2H3/t13-,14-,19-/m0/s1
Formula Weight 322.1681
Chemical Name or Material Quininone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.