CAS RN 84-31-1
CAS RN 84-31-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Quininone, TRC
CAS: 84-31-1 Molecular Formula: C20 H22 N2 O2 Molecular Weight (g/mol): 322.4 Synonym: (8α)-6'-Methoxycinchonan-9-one,Quininone (7CI,8CI),ICQ 15 IUPAC Name: [(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanone SMILES: COc1ccc2nccc(C(=O)[C@@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1