Learn More
Quinine N,N'-Dioxide (Quinine Di-N-Oxide), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Quinine N,N'-Dioxide (Quinine Di-N-Oxide)
Specifications
Specifications
| Chemical Name or Material | Quinine Di-N-oxide |
| CAS | 101655-92-9 |
| Analyte or Component Names | Quinine Di-N-oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Quinine N,N'-Dioxide (7CI), Quinine Di-N-Oxide, Cinchonan-9-ol, 6'-methoxy-, 1,1'-dioxide, (8alpha,9R)- |
| InChI Formula | InChI=1S/C20H24N2O4/c1-3-13-12-22(25)9-7-14(13)10-19(22)20(23)16-6-8-21(24)18-5-4-15(26-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19-,20+,22?/m0/s1 |
| SMILES | COc1ccc2c(c1)c(cc[n+]2[O-])[C@@H](O)[C@@H]3C[C@@H]4CC[N+]3([O-])C[C@@H]4C=C |
| IUPAC Name | (R)-[(2S,4S,5R)-5-ethenyl-1-oxido-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxy-1-oxidoquinolin-1-ium-4-yl)methanol |
| Show More |