Learn More
Quinazoline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 253-82-7 |
| Molecular Formula | C8H6N2 |
| MDL Number | MFCD00006712 |
| Quantity | 5 g |
| SMILES | C1=C2C(=NC=N1)C=CC=C2 |
| Molecular Weight (g/mol) | 130.15 |
| Percent Purity | 98% |
| Chemical Name or Material | Quinazoline |