Learn More
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Product Code. 12558855
Change view
Click to view available options
Standard Taper Joint:
7/16
Standard; 10/19
Standard; 12/21
Standard; 14/23
Standard; 19/26
Standard; 24/29
Standard; 29/32
Standard; 34/35
Standard; 45/40
Unit Size:
Pack of 10
Pack of 2
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
12558855 Standard; 10/19 Pack of 5
12508865 Standard; 24/29 Pack of 5
11349623 Standard; 34/35 Pack of 5
12568855 Standard; 19/26 Pack of 5
12508875 Standard; 29/32 Pack of 5
12495376 Standard; 45/40 Pack of 2
10155824 Standard; 14/23 Pack of 5
15557960 7/16 Pack of 10
15537960 Standard; 12/21 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12558855 Supplier Quickfit Supplier No. SB10
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. Azlon™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
  • Conforms to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chemical durability and high temperature performance
For Use With (Equipment) Quickfit™ jointed ware and other Glassware having standard ground Glass sockets
Standard Taper Joint Standard; 10/19
Material Borosilicate Glass
Product Type Hollow Stopper