Learn More
Quickfit™ Right Angle Tubing Adapter
Product Code. 12098361
Change view
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size unitSize
12098361 14/23 Each
11342035 19/26 Each
11352035 24/29 Each
11362035 29/32 Each
12008371 34/35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12098361 Supplier Quickfit Supplier No. MF10/1B/SC
Only null left
Add to basket
Add to basket
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
  • With right angle connection for attachment of rubber hose etc
  • Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Cone Size 14/23
For Use With (Equipment) Flexible tubing, Rubber hose
Includes Detachable Plastic screw thread connector
Material Borosilicate Glass
Product Type Right angle tubing adapter