Learn More
Quickfit™ Glass Büchner Funnel with Glass Sintered Disc
Product Code. 12425386
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
Diameter (Metric) Disc:
55mm
95mm
Cone Size:
19/26
24/29
29/32
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Disc Cone Size unitSize
12425386 125 mL 55mm 19/26 Each
12098473 125 mL 55mm 24/29 Each
12008483 500 mL 95mm 24/29 Each
15567960 125 mL 55mm 29/32 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12425386 Supplier Quickfit Supplier No. SF3A32
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
Used in place of conventional filter papers that would be destroyed by acids and alkalis. Quickfit™ Glass Büchner Funnel with Sintered Glass Disc is manufactured from chemical-resistant borosilicate glass. Integral ground glass cone on stem connects to Quickfit™ Büchner flasks and vessels.
  • Büchner funnel with glass sintered disc
  • Borosilicate glass construction for high resistance to chemical attack
  • Useful where conventional filter papers would be destroyed by acids and alkalis
  • Integral ground glass cone on stem for connecting to Quickfit™ Büchner flasks etc
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 125 mL
Material Borosilicate Glass
Cone Size 19/26
For Use With (Application) Bases or acids that can destroy conventional filter papers
Diameter (Metric) Disc 55mm
Includes Sintered glass disc, ground glass cone
Porosity 3
Pore Size 16 to 40μm
Product Type Buchner Funnel