Learn More
Quickfit™ Three-Neck Round-Bottom Glass Flask
Product Code. 12050653
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Socket Joint:
14/23 (Centre), 14/23 (side)
19/26 (Centre), 14/23 (side)
19/26 (Centre), 19/26 (side)
24/29 (Centre), 14/23 (side)
24/29 (Centre), 19/26 (side)
29/32
29/32 (Centre), 14/23 (side)
29/32 (Centre), 19/26 (side)
29/32 (Centre), 29/32 (side)
34/35 (Centre), 19/26 (side)
Unit Size:
Each
Pack of 5
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Socket Joint unitSize
12050653 250 mL 24/29 (Centre), 19/26 (side) Each
11710259 100 mL 24/29 (Centre), 14/23 (side) Each
10228054 1000 mL 24/29 (Centre), 19/26 (side) Each
12020643 100 mL 24/29 (Centre), 14/23 (side) Each
11301945 100 mL 19/26 (Centre), 14/23 (side) Each
12488966 100 mL 24/29 (Centre), 19/26 (side) Each
11371955 250 mL 19/26 (Centre), 19/26 (side) Each
12408996 250 mL 24/29 (Centre), 14/23 (side) Each
12622012 250 mL 29/32 (Centre), 19/26 (side) Each
12591738 25 mL 14/23 (Centre), 14/23 (side) Each
12030673 500 mL 24/29 (Centre), 19/26 (side) Each
11900895 500 mL 29/32 (Centre), 19/26 (side) Each
12493621 50 mL 14/23 (Centre), 14/23 (side) Each
12413631 50 mL 19/26 (Centre), 14/23 (side) Each
15557860 1000 mL 29/32 Pack of 5
15577860 1000 mL 29/32 (Centre), 29/32 (side) Pack of 5
15587860 1000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15547870 250 mL 29/32 (Centre), 14/23 (side) Pack of 5
15577870 2000 mL 24/29 (Centre), 19/26 (side) Pack of 5
15527880 2000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15597880 3000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15567890 50 mL 19/26 (Centre), 19/26 (side) Pack of 5
15587890 500 mL 24/29 (Centre), 14/23 (side) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 12050653 Leverantör Quickfit Leverantörsnummer FR250/3S/22A
Trusted Glassware Deals
Use promo code 28509
Visa alla kampanjer
Estimated Shipment: 02-09-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
  • Fitted with three Quickfit™ precision-ground sockets - One vertical centre neck - Two angled side necks
  • Quickfit™ ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
TRUSTED_SUSTAINABILITY
Specifikationer
Certifications/Compliance ISO 383
Capacity (Metric) 250 mL
Material Borosilicate Glass
Height (Metric) 130 mm
No. per Case 1
Socket Joint 24/29 (Centre), 19/26 (side)
Bottom Shape Round
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.