Learn More
Quickfit™ Borosilicate Glass Soxhlet Extractor
Product Code. 11371885
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
100 mL
200 mL
Standard Taper Joint Bottom:
24/29
29/32
34/35
40/38
50/42
Height (Metric):
175 mm
200 mm
205 mm
220 mm
240 mm
250 mm
Cone Size:
19/26
24/29
34/35
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Standard Taper Joint Bottom Height (Metric) Cone Size unitSize
11371885 100 mL 40/38 220 mm 24/29 Each
11381885 200 mL 50/42 250 mm 24/29 Each
12004793 100 mL 34/35 240 mm 34/35 Each
11331885 20 mL 24/29 175 mm 19/26 Each
11351885 60 mL 34/35 205 mm 24/29 Each
11341885 40 mL 29/32 200 mm 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11371885 Supplier Quickfit Supplier No. EX5/63
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
  • Conforms to BS 2071
  • For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
  • Wide choice of extractors to accommodate most sizes of sample and thimbles
  • Accurately formed concentric siphon tube for reliable performance
TRUSTED_SUSTAINABILITY
Specifications
Product Type Soxhlet Extractor
Capacity (Metric) 100 mL
Material Pyrex Borosilicate Glass
Cone Size 24/29
For Use With (Application) Ideal for extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
Certifications/Compliance BS 2071
Standard Taper Joint Bottom 40/38
Height (Metric) 220 mm