Learn More
Quickfit™ Borosilicate Glass Soxhlet Extractor
Product Code. 15597800
Change view
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15597800 60 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15597800 Supplier Quickfit Supplier No. EX5/55
  • ONLINE EXCLUSIVE PRICE
    685.00 EUR
    531.00 EUR / Pack of 5
    Save 154.00 EUR
    22% Off
Estimated Shipment: 08-04-2027
Conforms to BS 2071. Quickfit™ Borosilicate Glass Soxhlet Extractor removes soluble components or impurities from solids via repeated soaking/siphoning with solvent. A wide choice of extractors is available to accommodate most sizes of sample and thimbles. Accurately formed concentric siphon tube provides reliable performance.
  • Conforms to BS 2071
  • For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
  • Wide choice of extractors to accommodate most sizes of sample and thimbles
  • Accurately formed concentric siphon tube for reliable performance
Specifications
Product Type Soxhlet Extractor
Capacity (Metric) 60 mL
Material Pyrex Borosilicate Glass
Cone Size 34/35
For Use With (Application) Ideal for extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent
Certifications/Compliance BS 2071
Standard Taper Joint Bottom 34/35
Height (Metric) 205 mm