Learn More
- Home
- Products
- Flasks
- Boiling Flasks
- Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Product Code. 12498936
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
Height (Metric):
85 mm
95 mm
103 mm
105 mm
110 mm
115 mm
130 mm
135 mm
140 mm
145 mm
155 mm
170 mm
175 mm
187 mm
205 mm
215 mm
230 mm
260 mm
Socket Joint:
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
25 product options available for selection
| Product Code. | Capacity (Metric) | Height (Metric) | Socket Joint | unitSize |
|---|---|---|---|---|
| 12498936 | 1000 mL | 170 mm | 24/29 | Pack of 2 |
| 12498926 | 100 mL | 95 mm | 19/26 | Pack of 5 |
| 12418936 | 100 mL | 103 mm | 24/29 | Pack of 5 |
| 12428936 | 100 mL | 103 mm | 29/32 | Pack of 5 |
| 10243930 | 150 mL | 110 mm | 24/29 | Pack of 5 |
| 10697445 | 150 mL | 115 mm | 34/35 | Pack of 5 |
| 12428946 | 250 mL | 130 mm | 19/26 | Pack of 5 |
| 10309754 | 250 mL | 130 mm | 29/32 | Pack of 5 |
| 10831041 | 250 mL | 130 mm | 34/35 | Pack of 5 |
| 12509885 | 500 mL | 140 mm | 24/29 | Pack of 5 |
| 12418956 | 500 mL | 145 mm | 29/32 | Pack of 5 |
| 10851201 | 50 mL | 85 mm | 19/26 | Pack of 5 |
| 12661992 | 50 mL | 85 mm | 24/29 | Pack of 5 |
| 11980885 | 250 mL | 130 mm | 24/29 | Pack of 5 |
| 15527840 | 2000 mL | 215 mm | 34/35 | Pack of 5 |
| 12408946 | 1000 mL | 187 mm | 29/32 | Pack of 2 |
| 15567830 | 150 mL | 105 mm | 19/26 | Pack of 20 |
| 15577830 | 150 mL | 110 mm | 29/32 | Pack of 20 |
| 15517840 | 2000 mL | 230 mm | 29/32 | Pack of 5 |
| 15547840 | 500 mL | 135 mm | 19/26 | Pack of 20 |
| 15567840 | 500 mL | 155 mm | 34/35 | Pack of 20 |
| 15557830 | 100 mL | 103 mm | - | Pack of 10 |
| 15597830 | 1000 mL | 175 mm | 34/35 | Pack of 10 |
| 15507840 | 2000 mL | 205 mm | 24/29 | Pack of 5 |
| 15537840 | 4 L | 260 mm | - | Pack of 5 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
This item is not returnable.
View return policy
Product Code.
12498936
Supplier
Quickfit
Supplier No.
FF1L/3US
Trusted Glassware Deals
Use promo code 28509
Use promo code 28509
Estimated Shipment: 19-10-2026
Log in to see stock.
Log in to see stock.
Add to basket
83.70 EUR valid until 2026-08-30
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
BEST PRICE on Promo! Use promo code "28509" to get your promotional price.
Add to basket
This item is not returnable.
View return policy
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
- Complies with ISO 1773 and ISO 4797
- With highest quality Quickfit™ ground glass socket
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Flat base allows use with hotplates
Specifications
Specifications
| Certifications/Compliance | ISO 1773, ISO 4797 |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 170 mm |
| Neck Style | Short |
| Socket Joint | 24/29 |
| Bottom Shape | Flat |