Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Product Code. 12019652
Change view
Click to view available options
Length (Metric):
135 mm
260 mm
340 mm
380 mm
390 mm
400 mm
530 mm
535 mm
550 mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12019652 260 mm Each
11391845 135 mm Each
11301855 340 mm Each
12644911 380 mm Each
12039652 390 mm Each
11321855 535 mm Each
12575057 400 mm Each
12059652 550 mm Each
12049652 530 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12019652 Supplier Quickfit Supplier No. C1/11/SC
  • ONLINE EXCLUSIVE PRICE
    56.10 EUR
    48.63 EUR / Pack of 1
    Save 7.47 EUR
    13% Off
Estimated Shipment: 23-12-2026
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
Specifications
Certifications/Compliance DIN 12576
For Use With (Application) Suitable for distillation and reflux applications
Length (Metric) 260 mm
Standard Taper Joint No 14/23
Material Borosilicate Glass
Type Liebig Condenser