Learn More
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone
Product Code. 12070063
Change view
Click to view available options
Standard Taper Joint No:
34/35
40/38
50/42
Length (Metric) Jacket:
320 mm (Overall) 160 mm (Effective)
348 mm (Overall) 155 mm (Effective)
380 mm (Overall) 207 mm (Effective)
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
12070063 40/38 380 mm (Overall) 207 mm (Effective) Each
12060063 34/35 320 mm (Overall) 160 mm (Effective) Each
15537780 50/42 348 mm (Overall) 155 mm (Effective) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12070063 Supplier Quickfit Supplier No. CX3/06/SC
  • ONLINE EXCLUSIVE PRICE
    125.00 EUR
    106.93 EUR / Pack of 1
    Save 18.07 EUR
    14% Off
Estimated Shipment: 23-12-2026
Circulates coolant through a coil with a very large cooling surface. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone is ideal for use in the upright reflux mode where counter-current cooling is possible. Supplied with detachable plastic connectors for the attachment of rubber hose.
  • Coolant is circulated through a coil having a very large cooling surface 
  • Ideal for use in the upright reflux mode where counter-current cooling is possible
  • Supplied with detachable plastic connectors for the attachment of rubber hose
Specifications
For Use With (Application) Ideal for use in the upright reflux mode where counter-current cooling is possible
Length (Metric) Jacket 380 mm (Overall) 207 mm (Effective)
Standard Taper Joint No 40/38
Material Borosilicate Glass
Type Condenser Coil