Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Product Code. 15748127
Change view
Click to view available options
Capacity (Metric):
30 mL
80 mL
125 mL
500 mL
Porosity:
1
2
3
4
Diameter (Metric) Disc:
40 mm
40mm
60mm
95 mm
95mm
Disc: 30mm
Unit Size:
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Porosity Diameter (Metric) Disc Unit Size
15748127 500 mL 4 95mm Pack of 5
15728127 30 mL 4 Disc: 30mm Pack of 5
15528010 125 mL 4 60mm Pack of 5
15568000 80 mL 1 40mm Pack of 5
15738127 500 mL 2 95mm Pack of 5
15508010 125 mL 1 60mm Pack of 5
15598000 80 mL 4 40mm Pack of 5
15588000 80 mL 3 40 mm Pack of 5
15538010 500 mL 3 95 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15748127 Supplier Quickfit Supplier No. VB95/08M
  • ONLINE EXCLUSIVE PRICE
    760.00 EUR
    654.41 EUR / Pack of 5
    Save 105.59 EUR
    13% Off
Estimated Shipment: 09-04-2027
Useful where conventional filter papers would be destroyed by acids and alkalis
  • Manufactured from borosilicate glass for highest resistance to chemical attack
  • With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass
Height (Metric) 95 mm
Cone Size 29/32
Diameter (Metric) Disc 95mm
Porosity 4
Product Type Vacuum Filtration Funnel