missing translation for 'onlineSavingsMsg'
Learn More

Quercetin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30320457
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320457 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320457 Supplier Dr. Ehrenstorfer Supplier No. DREA16695000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quercetin
CAS 117-39-5
Analyte or Component Names Quercetin
Shipping Condition Room Temperature
Molecular Formula C15 H10 O7
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 4H-1-Benzopyran-4-one, 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-, Flavone, 3,3',4',5,7-pentahydroxy- (7CI,8CI), Flavone, 3,4',5,5',7-pentahydroxy- (6CI), 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-benzopyran-4-one, 3,3',4',5,7-Pentahydroxyflavone, 3,5,7,3',4'-Pentahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4-dihydroxyphenyl)-4H-chromen-4-one, 3'-Hydroxykaempferol, C.I. 75670, Corvitin, Cyanidelonon 1522, Korvitin, LDN 0052529, Lipoflavon, Meletin, NSC 57655, NSC 9219, Quercetin, Quercetine, Quercetol, Quertin, Quertine, Sophoretin, Xanthaurine
InChI Formula InChI=1S/C15H10O7/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6/h1-5,16-19,21H
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 302.24
Quantity 1 mL
Formula Weight 302.0427 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.