Learn More
Velp Scientifica™ Quartz Wool
Product Code. 15339574 Shop All Velp Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15339574 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15339574 Supplier Velp Scientifica™ Supplier No. A00000154
  • ONLINE EXCLUSIVE PRICE
    154.31 EUR
    145.00 EUR / 50g
    Save 9.31 EUR
    6% Off
Estimated Shipment: 23-11-2026
Velp Scientifica™ Quartz Wool for use with Dumas Nitrogen Analyzer NDA 701 and 702.
  • For use with NDA 701 and 702
Specifications
Type Quartz Wool
For Use With Dumas Nitrogen Analyzer
Quantity 50 g