Learn More
Hellma™ Quartz Suprasil Ultra-Micro Absorption Cells
Product Code. 10648972
Change view
Click to view available options
Capacity (Metric):
0.7 to 5 μL
100 μL
300 μL
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10648972 0.7 to 5 μL Each
10446491 100 μL Each
11756214 300 μL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10648972 Supplier Hellma™ Supplier No. 1052011540
  • ONLINE EXCLUSIVE PRICE
    427.00 EUR
    419.66 EUR / Pack of 1
    Save 7.34 EUR
    1% Off
Estimated Shipment: 22-10-2026
Specifications
Type Ultra-micro Absorption Cell
Capacity (Metric) 0.7 to 5 μL
Material Quartz Suprasil
Quantity 1 Each
Path Length 10 mm
Height (Metric) 45 mm
For Use With (Equipment) Spectrophotometer, Photometer
Width (Metric) 12.5 mm