Learn More
Applied Biosystems™ QuantStudio™ Absolute Q™ MAP16 Plate Kit and Master Mix
Product Code. 17427823 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
12 plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17427823 12 plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17427823 Supplier Applied Biosystems™ Supplier No. A53301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A key component of the QuantStudio Absolute Q Digital PCR System powered by the patented microfluidic array plate (MAP) technology, enabling consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV).
  • A key component of the QuantStudio Absolute Q Digital PCR System
  • System is powered by the patented microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV)
  • QuantStudio Absolute Q MAP16 consumable uses microinjection-molded plates to overcome common challenges of inconsistency and high dead volume
  • Benefits of this approach include precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed
  • Includes QuantStudio Absolute Q DNA Master Mix optimized for use with the QuantStudio Absolute Q Digital PCR System and Applied Biosystems digital PCR (dPCR) assays
  • 5X formulation enables analysis of higher sample volumes and delivers accurate quantification of DNA targets without using a standard curve
  • Unparalleled precision and accuracy of the Absolute Q system provides confidence in measurements across replicates and experiments
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
For Use With (Application) Digital PCR
Includes 12 MAP16 Plates, Gaskets, Isolation Buffer, and Absolute Q™ DNA Digital PCR Master Mix
Quantity 12 plates
compliance-icons compliance-icons compliance-icons
Product Identifier
  • QS ABS Q MAP16 PLATE MMIX KIT
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methyl-3-isothiazolone, Proteinase, tritirachium album serine, Ethanol, Isopropanol
For Research Use Only. Not for use in diagnostic procedures