Learn More
Macherey-Nagel™ Quantofix™ Test Strips
Product Code. 11807073 Shop All Macherey-Nagel Products
Change view
Click to view available options
Description:
LubriCheck
Peracetic Acid 500
Silver
Total Iron 1000
Range:
0, 1, 2, 3, 5, 7, 10g/L Ag
0, 15, 50, 75, 130, 200mmol/L KOH
0, 5, 20, 50, 100, 250, 500, 1000mg/L Fe2+/3+
0, 50, 100, 200, 300, 400, 500mg/L Peracetic Acid
Color Code:
White to Dark Red
Yellow to Blue
Yellow to Brown
Yellow to Green
For Use With (Application):
Determination of Peracetic Acid
Determination of Potassium Hydroxide
Determination of Total Iron 1000
Determination of silver
Unit Size:
Each
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Range Color Code For Use With (Application) Unit Size
11807073 Peracetic Acid 500 0, 50, 100, 200, 300, 400, 500mg/L Peracetic Acid Yellow to Green Determination of Peracetic Acid Pack of 100
15171136 Silver 0, 1, 2, 3, 5, 7, 10g/L Ag Yellow to Brown Determination of silver Each
10078222 LubriCheck 0, 15, 50, 75, 130, 200mmol/L KOH Yellow to Blue Determination of Potassium Hydroxide Pack of 100
12691515 Total Iron 1000 0, 5, 20, 50, 100, 250, 500, 1000mg/L Fe2+/3+ White to Dark Red Determination of Total Iron 1000 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11807073 Supplier Macherey-Nagel™ Supplier No. 91341
  • ONLINE EXCLUSIVE PRICE
    43.70 EUR
    36.65 EUR / Pack of 100
    Save 7.05 EUR
    16% Off
 Restricted Item
In Stock
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Quantofix™ test strips meet all requirements of a modern rapid test. Analytical professionals as well as occasional testers appreciate the test strips for the fast and easy analysis directly at the point of interest. Often, these tests are used to quickly check whether important parameters are in the desired range. They deliver an immediate result and thus enable a fast response.
  • Rapid
    • Just Dip & Read
    • Results within seconds
    • Always ready for use
  • Easy
    • No calibration
    • No maintenance
    • No accessories
  • Reliable
    • Desiccant in the stopper for optimal protection of the strips against humidity
    • Color chart confirmed with traceable standards
    • Automatic evaluation with Quantofix™ Relax for safe documentation
Specifications
Certifications/Compliance CE for disinfection/medical products
Color Code Yellow to Green
Description Peracetic Acid 500
Detectable Analytes Peracetic Acid
Format Semi Quantitative Test Strips
Type Redox Reaction
For Use With (Application) Determination of Peracetic Acid
Quantity 50 - 500mg/L Peracetic Acid
Range 0, 50, 100, 200, 300, 400, 500mg/L Peracetic Acid
Test Time 30 sec.