Learn More
Macherey-Nagel™ Quantofix™ Nitrate and Nitrite Test Strip Kits
Product Code. 10384071 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
White to Red/Violet
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10384071 White to Red/Violet Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10384071 Supplier Macherey-Nagel™ Supplier No. 91313
  • ONLINE EXCLUSIVE PRICE
    40.60 EUR
    35.70 EUR / Pack of 100
    Save 4.90 EUR
    12% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Allows for the quick and easy determination of nitrate and nitrite in solutions. Macherey Nagel™ Quantofix™ Nitrate and Nitrite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Allows for the quick and easy determination of nitrate and nitrite in solutions. Macherey Nagel™ Quantofix™ Nitrate and Nitrite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Rapid: provides reliable results within one minute
  • Easy dip-and-read procedure
  • Excellent for use in the food and farming industries
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C
Especificaciones
Color Indicator White to Red/Violet
Format Strip
For Use With (Application) Determination of Nitrate and Nitrite
Graduations 0, 10, 25, 50, 100, 250, 500mg/L NO3- (Nitrate), 0, 1, 5, 10, 20, 40, 80mg/L NO2- (Nitrite)
Quantity 100 Pack
Storage Requirements 30°C
Test Time 1 min.
Type Semi-qualitative Test Strip
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.