Learn More
R&D Systems™ Quantikine Immunoassay Control Set 665 for Human Uteroglobin
Product Code. 30001940
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001940 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001940 Supplier R&D Systems™ Supplier No. QC181
  • ONLINE EXCLUSIVE PRICE
    221.93 EUR
    198.45 EUR / Pack of 1
    Save 23.48 EUR
    10% Off
 Restricted Item
Estimated Shipment: 03-12-2026
For use with catalog number DUGB00
These lyophilized controls are designed for use as quantitative controls for the determination of Uteroglobin concentrations in biological fluids.
  • Product Type: Immunoassay Control Set
  • Analyte: Human Uteroglobin
  • Matrix: Diluted porcine serum with preservatives
  • Format: Lyophilized
  • Intended Use: For research use only
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Uteroglobin/SCGB1A1
Type Immunoassay Control Set
Gene ID (Entrez) 7356
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized