Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1180 for Human Pro-MMP-13
Description
Quantikine™ Immunoassay Control Set 1180 for Human Pro-MMP-13 is designed for use as quantitative controls for the determination of cytokine concentrations in biological fluids. These controls are prepared in a buffered protein solution with preservatives and contain recombinant human cytokines at low, medium, and high concentrations.
- Species Reactivity: Human
- Form: Lyophilized
- Applications: ELISA control
- Storage: Unreconstituted controls should be stored at 2°C to 8°C. Reconstituted controls may be stable when stored at <-20°C, depending on the analyte.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | MMP-13 |
| Type | Immunoassay Control Set |
| Gene ID (Entrez) | 4322 |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
| Source | Human |
| Form | Lyophilized |