Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1178 for Human COMP
Product Code. 30001976
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001976 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30001976 Supplier R&D Systems™ Supplier No. QC66
  • ONLINE EXCLUSIVE PRICE
    210.00 EUR
    198.45 EUR / Pack of 1
    Save 11.55 EUR
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 06-10-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
For use with catalog number DCMP0
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes COMP (Thrombospondin-5)
Gene ID (Entrez) 1311
Intended Use Control
Quantity 1 Each
Source Human