Learn More
Kolmi Hopen™ Qualitative Mask Fit Test Kit
Product Code. 17730229 Shop All Medicom Products
Change view
Click to view available options
Unit Size:
8 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17730229 8 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17730229 Supplier Kolmi Hopen™ Supplier No. QLFTK02
  • ONLINE EXCLUSIVE PRICE
    3055.00 EUR
    2756.59 EUR / 8 sets
    Save 298.41 EUR
    9% Off
Estimated Shipment: 10-12-2026
This kit gives everything that a fit tester needs, wherever they are conducting mask fitting tests.
Dedicated to Fit Testers, these handy-sized A4 packs are perfect for use when carrying out Fit Tests across the NHS.
  • Many items are printed on easy-clean material for multiple uses
  • Includes 3 mask sizing guides, 6 donning/doffing posters, 35 evaluation cards with permanent marker, 10 wearer guides, and fit test guide
Specifications
Includes Sizing Guides, Posters, Evaluation Cards, Wearer and Fit Test Guides