Learn More
Invitrogen™ Qualification Add-on For FL1000 Imaging System
Product Code. 15746335
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15746335 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15746335 Supplier Invitrogen™ Supplier No. ZGQUSCIBRIGHTFL
 Restricted Item
Estimated Shipment: 11-11-2026
Designed to help balance business and laboratory requirements and help comply with global standards, country-specific regulations, and other laboratory site requirements.
  • Verify instrument performance and minimize downtime with IQ/OQ services (Installation Qualification and Operation Qualification)
  • Complete with audit ready documentation, factory-trained and experienced engineers will conduct IQ/OQ services
  • Qualification services help ensure instruments are installed and operating according to manufacturer's specifications
  • Helps move into production faster
  • IQ/OQ recommended: At installation , after a move
  • OQ recommended: After planned maintenance, After major repair, After additions, changes, or upgrades to the system, For a scheduled OQ site requirement
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) iBright FL1000 Imaging System
Product Type Qualification Add-on
For Use With (Application) Warranty for Western Blotting instrumentation
Includes Audit-ready documentation, Order and system verification, Documentation verification, Installation verification, Software verification, Hardware operation verification, Component verification, Service coverage verification, Customer training verification