Learn More
QNZ, MedChemExpress
Product Code. 16418527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418527 5 mg 5mg
16412027 50 mg 50mg
16405878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16418527 Supplier Medchem Express Supplier No. HY13812
MedChemExpress QNZ (EVP4593) shows strong inhibitory effects on NF-κB transcriptional activation and TNF-α production with IC50s of 11 and 7 nM, respectively. QNZ (EVP4593) is a neuroprotective inhibitor of SOC channel.
QNZ (EVP4593) shows strong inhibitory effects on NF-κB transcriptional activation and TNF-α production with IC50s of 11 and 7 nM, respectively. QNZ (EVP4593) is a neuroprotective inhibitor of SOC channel.
Specifications
Chemical Name or Material QNZ
CAS 545380-34-5
Color Green
Purity Grade Notes Research
Molecular Formula C22H20N4O
Quantity 5 mg
Synonym EVP4593
Solubility Information DMSO : ≥ 37 mg/mL (103.81 mM)
SMILES NC1=CC2=C(NCCC3=CC=C(OC4=CC=CC=C4)C=C3)N=CN=C2C=C1
Molecular Weight (g/mol) 356.42
Formula Weight 356.42
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation