missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyruvic Acid Sodium Salt, Millipore™
Description
Zwitterionic buffer substanceMES is used as a zwitterionic buffer substance in the preparation of media for the culture of microorganisms and plant cells. MES is a very convenient buffer substance due to its biological inertness.
Specifications
Specifications
| Melting Point | 236.8°C |
| Quantity | 1 kg |
| MDL Number | MFCD00002586 |
| Synonym | Pyruvic acid sodium salt, Sodium pyruvate, Pyruvate, Pyruvic acid sodium salt |
| Molecular Weight (g/mol) | 110.04 |
| Grade | Research |
| Density | 1.78 g/cm3 at 20°C |
| Chemical Name or Material | Pyruvic Acid Sodium Salt |
Safety and Handling
Product Identifier
- Pyruvic acid sodium salt for biochemistry
- Warning
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash skin thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Pyruvic acid sodiumsalt
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?