missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid, Sodium Salt, ∽99%, Reagent Grade, MP Biomedicals™

Product Code. 11496322 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496322 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496322 Supplier MP Biomedicals Supplier No. 0210292690
  • ONLINE EXCLUSIVE PRICE
    550.00 EUR
    523.95 EUR / 500g
    Save 26.05 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium pyruvate is utilized as a component in culture broth and media

Specifications

CAS 113-24-6
Color White
Assay Percent Range Approx. 99%
Molecular Formula C3H3NaO3
MDL Number MFCD00002586
Quantity 500 g
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
SMILES CC(=O)C(=O)[O-].[Na+]
IUPAC Name sodium;2-oxopropanoate
Molecular Weight (g/mol) 110.044
PubChem CID 23662274
ChEBI CHEBI:50144
Formula Weight 110.04
Grade Reagent
Physical Form Powder
Chemical Name or Material Pyruvic Acid, Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.